missing translation for 'onlineSavingsMsg'
Learn More

Dkk-1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1096

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436734

  • £390.01 / 100µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dkk-1 Polyclonal antibody AF1096 Specifically detects Dkk-1 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

Dkk-1
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Knockout Validated
O94907
dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Dkk-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 75% cross-reactivity with recombinant mouse (rm) Dkk-1 50% cross-reactivity with recombinant rat Dkk-1 is observed, and less than 1% cross-reactivity with recombinant human (rh) Dkk-2, rhDkk-3, rhDkk-4, and rmDkk-3 is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction, Immunoassay
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Affinity Purified
DKK1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907
Antigen Affinity-purified
RUO
Primary
22943
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only