missing translation for 'onlineSavingsMsg'
Learn More

Desmoglein-3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1720

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437114

  • £314.17 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Desmoglein-3 Polyclonal antibody AF1720 Specifically detects Desmoglein-3 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Desmoglein-3
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
P32926
Cadherin family member 6, CDHF6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, Desmoglein3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, DSG3, pemphigus vulgaris antigen, PVA
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Desmoglein-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Desmoglein-1 and rhDesmoglein-2 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DSG3
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-3 Glu50-Arg615 Accession # P32926
Antigen Affinity-purified
RUO
Primary
1830
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only