missing translation for 'onlineSavingsMsg'
Learn More

Desmocollin-3 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4630

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448824

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Desmocollin-3 Polyclonal antibody AF4630 Specifically detects Desmocollin-3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Desmocollin-3
LYOPH
Western Blot 0.1 ug/mL
Q14574
Cadherin family member 3, CDHF3, CDHF3HT-CP, desmocollin 3, Desmocollin3, desmocollin-4, DSC, DSC1, DSC2, DSC3, DSC4, DSC4desmocollin-3, HT-CP
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Desmocollin-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Desmocollin-2 is observed and less than 1% cross-reactivity with rhDesmocollin-1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DSC3
Mouse myeloma cell line NS0-derived recombinant human Desmocollin-3 Arg136-Lys690 Accession # Q14574
Antigen Affinity-purified
RUO
Primary
1825
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only