missing translation for 'onlineSavingsMsg'
Learn More

DAN Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF955

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440355

  • £362.60 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DAN Polyclonal antibody AF955 Specifically detects DAN in Human samples. It is validated for Blocking (Receptor-ligand), Immunohistochemistry, Western Blotting
Specifications

Specifications

DAN/NBL1
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction
Q5U0N4
DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human DAN in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) DAN is observed and less than 1% cross-reactivity with recombinant chicken Caronte, rmGremlin, and recombinant human Cerberus 1 is observed.
Human
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NBL1
Mouse myeloma cell line NS0-derived recombinant human DAN Ala17-Asp180 (Ala36Arg) Accession # Q5U0N4
Immunogen affinity purified
RUO
Primary
4681
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only