missing translation for 'onlineSavingsMsg'
Learn More

Cystatin S Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1296

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486864

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Cystatin S Polyclonal antibody AF1296 Specifically detects Cystatin S in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cystatin S
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P01036
CST4, cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Cystatin S in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) Cystatin SA and rhCystatin SN is observed and 5% cross-reactivity with rhCystatin C and rhCystatin D is observed, and less than 1% cross-reactivity with rhCystatin A, rhCystatin B, rhCystatin E/M, rhFetuin A, and rhFetuin B is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CST4
Mouse myeloma cell line NS0-derived recombinant human Cystatin S, Ser21-Ala141, Accession # P01036
Immunogen affinity purified
RUO
Primary
1472
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only