missing translation for 'onlineSavingsMsg'
Learn More

Cystatin D Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1202

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486804

  • £347.17 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Cystatin D Polyclonal antibody AF1202 Specifically detects Cystatin D in Human samples. It is validated for Neutralization, Western Blotting
Specifications

Specifications

Cystatin D/Cst5
LYOPH
Western Blot 0.1 ug/mL, Neutralization 13 ug/mL
CAA49838
CST5, CSTD, cystatin 5, cystatin D, Cystatin-5, cystatin-D, cysteine-proteinase inhibitor, MGC71922
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Cystatin D in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) Cystatin C, rhCystatin SN, and rhCystatin S is observed and less than 1% cross-reactivity with rhCystatin E/M and rhFetuin A is observed.
Human
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CST5
Mouse myeloma cell line NS0-derived recombinant human Cystatin D, Thr29-Val142, Accession # CAA49838
Antigen Affinity-purified
RUO
Primary
1473
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only