missing translation for 'onlineSavingsMsg'
Learn More

CXCL17/VCC-1 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4207

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448614

  • £349.86 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CXCL17/VCC-1 Polyclonal antibody AF4207 Specifically detects CXCL17/VCC-1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CXCL17/VCC-1
LYOPH
Western Blot 0.1 ug/mL
Q6UXB2
Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, CXCL17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human DMC in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse DMC is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CXCL17
E. coli-derived recombinant human DMC Leu24-Leu119 Accession # Q6UXB2
Immunogen affinity purified
RUO
Primary
284340
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only