missing translation for 'onlineSavingsMsg'
Learn More

CXCL14/BRAK Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF866

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440285

  • £314.17 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CXCL14/BRAK Polyclonal antibody AF866 Specifically detects CXCL14/BRAK in Human samples. It is validated for Western Blotting
Specifications

Specifications

CXCL14/BRAK
LYOPH
Western Blot 0.1 ug/mL
O95715
BMAC, Bolekine, BRAK, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, CXC-X3, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CXCL14/BRAK in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse CXCL14/BRAK is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CXCL14
E. coli-derived recombinant human CXCL14/BRAK (R&D Systems, Catalog # 866-CX) Ser23-Glu99 Accession # O95715
Antigen Affinity-purified
RUO
Primary
9547
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only