missing translation for 'onlineSavingsMsg'
Learn More

CXADR Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3336

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448114

  • £398.37 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CXADR Polyclonal antibody AF3336 Specifically detects CXADR in Human samples. It is validated for Western Blotting
Specifications

Specifications

CXADR
LYOPH
Western Blot 0.1 ug/mL
P78310
CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3 BP, CVB3-binding protein, hCAR, HCVADR
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CXADR in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse CXADR is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CXADR
Mouse myeloma cell line NS0-derived recombinant human CXADR Leu20-Gly237 Accession # P78310
Immunogen affinity purified
RUO
Primary
1525
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only