Learn More
CSL Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF4079
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428544
  • £344.84 / 100µg
Estimated Shipment
17-09-2020
Description
Description
CSL Polyclonal antibody AF4079 Specifically detects CSL in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
csl/RBPJK
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Q06330
CBF1, CBF-1, csl, H-2K binding factor-2, IGKJRB1, IGKJRB1RBPSUHCBF1, IGKJRBRBP-J kappa, immunoglobulin kappa J region recombination signal binding protein 1, J kappa-recombination signal-binding protein, KBF2, RBPJ, RBP-JK, RBPJKMGC61669, RBP-Jrecombining binding protein suppressor of hairless (Drosophila), RBPSUH, recombination signal binding protein for immunoglobulin kappa J region, recombining binding protein suppressor of hairless, Renal carcinoma antigen NY-REN-30, SUH, suppressor of hairless homolog
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CSL in direct ELISAs and Western blots.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RBPJ
E. coli-derived recombinant human CSL Gly208-Ser500 Accession # Q06330
Immunogen affinity purified
RUO
Primary
3516
Reconstitute at 0.2 mg/mL in sterile PBS.