missing translation for 'onlineSavingsMsg'
Learn More

CRISP-3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2397

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437534

  • £314.17 / 100µg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CRISP-3 Polyclonal antibody AF2397 Specifically detects CRISP-3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CRISP-3
LYOPH
Western Blot 0.1 ug/mL
CAA63984
Aeg2, CRISP3, CRISP-3MGC126588, CRS3, cysteine-rich secretory protein 3, cysteine-rich secretory protein-3, dJ442L6.3, SGP28, SGP28Aeg2, specific granule protein (28 kDa), Specific granule protein of 28 kDa
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CRISP-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CRISP-2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CRISP3
Mouse myeloma cell line NS0-derived recombinant human CRISP-3 Asn21-Tyr245 (Ser134Ala) Accession # CAA63984
Immunogen affinity purified
RUO
Primary
10321
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only