Learn More
CRIM1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1917
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487224
  • £377.53 / 100µg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
CRIM1 Polyclonal antibody AF1917 Specifically detects CRIM1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
CRIM1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human CRIM1, Leu35-Ser939, Accession No. Q9NZV1
100μg
Primary
51232
Unconjugated
Q9NZV1
Goat