missing translation for 'onlineSavingsMsg'
Learn More

CRELD2 alpha Isoform Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3555

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458234

  • £389.41 / 100µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

CRELD2 alpha Isoform Polyclonal antibody AF3555 Specifically detects CRELD2 alpha Isoform in Human samples. It is validated for Western Blotting
Specifications

Specifications

CRELD2
LYOPH
Western Blot 0.1 ug/mL
AAH50675
cysteine-rich with EGF-like domain protein 2, cysteine-rich with EGF-like domains 2, DKFZp667O055, MGC11256
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CRELD2 alpha isoform in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) CRELD2 is observed and less than 1% cross-reactivity with recombinant human CRELD1 and rmCRELD1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CRELD2
Mouse myeloma cell line NS0-derived recombinant human CRELD2 alpha Isoform Ala21-Leu353 Accession # AAH50675
Immunogen affinity purified
RUO
Primary
79174
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only