Learn More
CRELD1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3128
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13457994
  • £314.17 / 100µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
CRELD1 Polyclonal antibody AF3128 Specifically detects CRELD1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
CRELD1
LYOPH
Western Blot 0.1 ug/mL
Q96HD1
atrioventricular septal defect 2, AVSD2, CIRRIN, cysteine-rich with EGF-like domain protein 1, cysteine-rich with EGF-like domains 1, DKFZp566D213
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CRELD1 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CRELD1
Mouse myeloma cell line NS0-derived recombinant human CRELD1 isoform 1 Gln30-Glu362 Accession # Q96HD1
Immunogen affinity purified
RUO
Primary
78987
Reconstitute at 0.2 mg/mL in sterile PBS.