missing translation for 'onlineSavingsMsg'
Learn More

Cornulin Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3607

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458264

  • £377.53 / 100µg


Estimated Shipment
03-12-2018
Add to basket

Description

Description

Cornulin Polyclonal antibody AF3607 Specifically detects Cornulin in Human samples. It is validated for Western Blotting
Specifications

Specifications

Cornulin
LYOPH
Western Blot 0.1 ug/mL
Q9UBG3
53 kDa squamous epithelial-induced stress protein, 58 kDa heat shock protein, C1orf10, chromosome 1 open reading frame 10, cornulin, CRNN, DRC1, Gm1015, PDRC1, PDRC1,53 kDa putative calcium-binding protein, SEP53, SEP53C1orf10, Squamous epithelial heat shock protein 53, Tumor-related protein
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Cornulin in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CRNN
E. coli-derived recombinant human Cornulin, Met1-Ser140, Accession # Q9UBG3
Immunogen affinity purified
RUO
Primary
49860
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only