missing translation for 'onlineSavingsMsg'
Learn More

Contactin-5 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3030

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467934

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Contactin-5 Polyclonal antibody AF3030 Specifically detects Contactin-5 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Contactin-5
LYOPH
Western Blot 0.1 ug/mL
O94779
CNTN5, contactin 5, contactin-5, Contactin5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Contactin-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Contactin-2 and rhContactin-3 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CNTN5
Mouse myeloma cell line NS0-derived recombinant human Contactin-5 Glu19-Gln1059 Accession # O94779
Antigen Affinity-purified
RUO
Primary
53942
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documenti

For Research Use Only