missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor H-related 1/CFHR1 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4247

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438634

  • £298.33 / 100µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Complement Factor H-related 1/CFHR1 Polyclonal antibody AF4247 Specifically detects Complement Factor H-related 1/CFHR1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Complement Factor H-related 1/CFHR1/CFHL1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q03591
BTA, CFHL1FHR-1, CFHLCFHR1P, CFHR1, complement factor H-related 1, Complement Factor Hrelated 1, complement factor H-related 1 pseudogene, complement factor H-related protein 1, FHR1, FHR1CFHL1P, H factor (complement)-like 1, H factor (complement)-like 2, H factor-like protein 1, H36-1, H36-2, H-factor-like 1, HFL1H36, HFL2, MGC104329
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Complement Factor H-related 1/CFHR1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) FHR2 is observed and approximately 15% cross-reactivity with rhFHR5 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CFHR1
Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 1/CFHR1 Glu19-Ala328 Accession # Q03591
Immunogen affinity purified
RUO
Primary
3078
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only