missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor B Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2739

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437754

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 18-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Complement Factor B Polyclonal antibody AF2739 Specifically detects Complement Factor B in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Complement Factor B
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
AAA16820
BF, BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, CFAB, CFB, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBG, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Complement Factor B in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human Complement Component C2 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CFB
Mouse myeloma cell line NS0-derived recombinant human Complement Factor B Thr26-Leu764 (Gln34Arg) Accession # AAA16820
Antigen Affinity-purified
RUO
Primary
629
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only