missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C1rLP Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3676

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458304

  • £322.09 / 100µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Complement Component C1rLP Polyclonal antibody AF3676 Specifically detects Complement Component C1rLP in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Complement Component C1rLP
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
AAI56781
C1RL, C1RL1, C1RL1complement C1r subcomponent-like protein, C1r-like protein, C1r-like serine protease analog protein, C1r-LPcomplement C1r-like proteinase, C1RLPEC 3.4.21.-, CLSPa, complement component 1, r subcomponent-like, EC 3.4.21, EC 3.4.21.41
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Complement Component C1rLP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse C1rLP is observed and less than 5% cross-reactivity with recombinant human (rh) C1R, rhC1S, and rhMASP-3 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
C1RL
Mouse myeloma cell line NS0-derived recombinant human Complement Component C1rLP Ser41-Asn487 Accession # AAI56781
Immunogen affinity purified
RUO
Primary
51279
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only