missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C1r Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1807

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467164

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Complement Component C1r Polyclonal antibody AF1807 Specifically detects Complement Component C1r in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Complement Component C1r
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
NP_001724
C1r, complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Complement Component C1r in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Complement Component C1s is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
C1R
Mouse myeloma cell line NS0-derived recombinant human Complement Component C1r Ser18-Asp705 Accession # NP_001724
Antigen Affinity-purified
RUO
Primary
715
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only