missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COCO Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand: RD Systems (Bio-Techne) AF3047
Code : Z1
Additional Details : Weight : 0.00010kg
Description
COCO Polyclonal antibody AF3047 Specifically detects COCO in Human samples. It is validated for Western BlottingSpecifications
COCO/DAND5 | |
LYOPH | |
Western Blot 0.1 ug/mL | |
NP_689867 | |
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
Goat | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human COCO in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant mouse COCO is observed. | |
Human |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
DAND5 | |
E. coli-derived recombinant human COCO (R&D Systems, Catalog # 3047-CC) Arg23-Ala189 Accession # NP_689867 | |
Immunogen affinity purified | |
RUO | |
Primary | |
199699 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
For Research Use Only