missing translation for 'onlineSavingsMsg'
Learn More

Coagulation Factor III/Tissue Factor Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2339

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437494

  • £389.41 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Coagulation Factor III/Tissue Factor Polyclonal antibody AF2339 Specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications

Specifications

Coagulation Factor III/Tissue Factor
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, CyTOF-ready
P13726
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse Coagulation Factor III/Tissue Factor is observed.
Human
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
F3
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III/Tissue Factor Gly34-Glu251 Accession # P13726
Immunogen affinity purified
RUO
Primary
2152
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only