missing translation for 'onlineSavingsMsg'
Learn More

CNTF Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AB-257-NA

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13446574

  • £360.15 / 1mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:E. coli-derived recombinant human CNTF, Met1-Met200, Accession No. P26441

Applications:

Neutralization, Western Blotting

Specifications

Specifications

CNTF
LYOPH
Western Blot 1 ug/mL, Neutralization 15-30 ug/mL
P26441
ciliary neurotrophic factor, HCNTF
Goat
IgG
1 mg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CNTF in direct ELISAs and Western blots.
Human
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Affinity Purified
CNTF
E. coli-derived recombinant human CNTF Met1-Met200 Accession # P26441
Protein A or G purified
RUO
Primary
1270
Reconstitute at 1 mg/mL in sterile PBS.
Videos
Documents