missing translation for 'onlineSavingsMsg'
Learn More

CNP Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3127

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447994

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CNP Polyclonal antibody AF3127 Specifically detects CNP in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications

Specifications

CNP/NPPC
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
P23582
CNP, CNP2, C-type natriuretic peptide, natriuretic peptide C, natriuretic peptide precursor C, NPPC
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CNP in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human ANP is observed.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NPPC
E. coli-derived recombinant human CNP Lys24-Cys126 Accession # P23582
Antigen Affinity-purified
RUO
Primary
4880
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only