Learn More
Clusterin-like 1/CLUL1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3185
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488024
  • £382.00 / 100µg
Estimated Shipment
02-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Clusterin-like 1/CLUL1 Polyclonal antibody AF3185 Specifically detects Clusterin-like 1/CLUL1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Clusterin-like 1/CLUL1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human Clusterin-like 1/CLUL1, Cys20-Ala442, Accession No. Q15846
100μg
Primary
27098
Unconjugated
Q15846
Goat