missing translation for 'onlineSavingsMsg'
Learn More

CLEC3B/Tetranectin Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF5170

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459124

  • £314.17 / 100µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

CLEC3B/Tetranectin Polyclonal antibody AF5170 Specifically detects CLEC3B/Tetranectin in Human samples. It is validated for Western Blotting
Specifications

Specifications

CLEC3B/Tetranectin
LYOPH
Western Blot 0.1 ug/mL
P05452
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, TETN, Tetranectin, tetranectin (plasminogen binding protein), TNA, TNAtetranectin (plasminogen-binding protein), TNtetranectin
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CLEC3B/Tetranectin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) CLEC1, rhCLEC2, rhCLEC10A, and rhCLEC14A is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CLEC3B
Mouse myeloma cell line NS0-derived recombinant human CLEC3B/Tetranectin Glu22-Val202 Accession # P05452
Immunogen affinity purified
RUO
Primary
7123
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only