Learn More
Chondrolectin Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2576
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13437654
  • £362.60 / 100µg
Estimated Shipment date
from Supplier 09-07-2020
Description
Description
Chondrolectin Polyclonal antibody AF2576 Specifically detects Chondrolectin in Human samples. It is validated for Western Blotting
Specifications
Specifications
Chondrolectin
LYOPH
Western Blot 0.1 ug/mL
Q9H9P2
C21orf68, CHODL, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Chondrolectin in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CHODL
Mouse myeloma cell line NS0-derived recombinant human Chondrolectin Arg22-Asn216 Accession # Q9H9P2
Immunogen affinity purified
RUO
Primary
140578
Reconstitute at 0.2 mg/mL in sterile PBS.