missing translation for 'onlineSavingsMsg'
Learn More

CHL-1/L1CAM-2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2126

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437364

  • £412.92 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CHL-1/L1CAM-2 Polyclonal antibody AF2126 Specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CHL-1/L1CAM-2
LYOPH
Western Blot 0.1 ug/mL
EAW63869
CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse CHL-1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CHL1
Mouse myeloma cell line NS0-derived recombinant human CHL-1/L1CAM-2 Ile25-Gln1096 Accession # EAW63869
Immunogen affinity purified
RUO
Primary
10752
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only