Learn More
CD97 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2529
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487614
  • £382.00 / 100µg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
CD97 Polyclonal antibody AF2529 Specifically detects CD97 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
CD97
Unconjugated
NP_001775
Goat
IgG
RUO
Polyclonal
Human
CyTOFþFlow CytometryþWestern Blotting
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human CD97, Gln21-Gln398, Accession # NP_001775
100μg
Primary
976