Learn More
CD68/SR-D1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2040
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13447304
  • £433.65 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
CD68/SR-D1 Polyclonal antibody AF2040 Specifically detects CD68/SR-D1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
CD68/SR-D1
LYOPH
Western Blot 0.1 ug/mL
AAB25811
CD68, CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, gp110, Macrosialin, SCARD1, scavenger receptor class D, member 1, SRD1, SR-D1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CD68/SR-D1 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CD68
Mouse myeloma cell line NS0-derived recombinant human CD68/SR-D1 Asn22-Ile320 Accession # AAB25811
Immunogen affinity purified
RUO
Primary
968
Reconstitute at 0.2 mg/mL in sterile PBS.