missing translation for 'onlineSavingsMsg'
Learn More

CD300c/LMIR2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3256

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438064

  • £362.60 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD300c/LMIR2 Polyclonal antibody AF3256 Specifically detects CD300c/LMIR2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CD300c/LMIR2
LYOPH
Western Blot 0.1 ug/mL
Q08708
CD300c antigenCMRF35 antigen, CD300c molecule, CLM4, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR, LMIR2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CD300c/LMIR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) LMIR1 is observed and less than 1% cross-reactivity with rhLMIR3, rhLMIR4, rhLMIR5, and rhLMIR6 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CD300C
Mouse myeloma cell line NS0-derived recombinant human CD300c/LMIR2 Met29-Arg183 Accession # Q08708
Immunogen affinity purified
RUO
Primary
10871
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only