Learn More
CD229/SLAMF3 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1898
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13467214
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
CD229/SLAMF3 Polyclonal antibody AF1898 Specifically detects CD229/SLAMF3 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
CD229/SLAMF3/Lymphocyte Antigen 9
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9HBG7
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, Ly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CD229/SLAMF3 in direct ELISAs and Western blots.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LY9
Mouse myeloma cell line NS0-derived recombinant human CD229/SLAMF3 Lys48-Lys454 Accession # Q9HBG7
Immunogen affinity purified
RUO
Primary
4063
Reconstitute at 0.2 mg/mL in sterile PBS.