missing translation for 'onlineSavingsMsg'
Learn More

CCL18/PARC Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF394

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438454

  • £389.41 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CCL18/PARC Polyclonal antibody AF394 Specifically detects CCL18/PARC in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blotting
Specifications

Specifications

CCL18/PARC
LYOPH
Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P55774
Alternative macrophage activation-associated CC chemokine 1, AMAC-1, AMAC1CC chemokine ligand 18, AMAC-1Small-inducible cytokine A18, chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated), CKb7, DC-CK1, DC-CK1CC chemokine PARC, DCCK1Macrophage inflammatory protein 4, Dendritic cell chemokine 1, MIP4, MIP-4, MIP-4C-C motif chemokine 18, PARC, PARCPulmonary and activation-regulated chemokine, SCYA18chemokine (C-C), dendritic, small inducible cytokine A18, small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary andactivation-regulated
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CCL18/PARC in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) MIP-1 alpha and recombinant mouse MIP-1 alpha is observed and less than 10% cross-reactivity with rhMPIF-1, recombinant viral (rv) MIP-I, rvMIP-II, rhHCC-1, and rhMIP-1 delta is observed.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CCL18
E. coli-derived recombinant human CCL18/PARC Ala21-Ala89 Accession # P55774
Antigen Affinity-purified
RUO
Primary
6362
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only