Learn More
Carnosine Dipeptidase 1/CNDP1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2489
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13447594
  • £360.15 / 100µg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Carnosine Dipeptidase 1/CNDP1 Polyclonal antibody AF2489 Specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
Carnosine Dipeptidase 1/CNDP1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL
Q96KN2
beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CNDP1, CPGL2, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Carnosine Dipeptidase 1/CNDP1 in direct ELISAs and Western blots.
Human
Western Blot, Immunoprecipitation, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CNDP1
Mouse myeloma cell line NS0-derived recombinant human Carnosine Dipeptidase 1/CNDP1 Pro28-His507 Accession # Q96KN2
Immunogen affinity purified
RUO
Primary
84735
Reconstitute at 0.2 mg/mL in sterile PBS.