missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase X1/CPXM1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2919

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457864

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Carboxypeptidase X1/CPXM1 Polyclonal antibody AF2919 Specifically detects Carboxypeptidase X1/CPXM1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Carboxypeptidase X1/CPXM1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q96SM3
carboxypeptidase X (M14 family), carboxypeptidase X (M14 family), member 1, CPX1, CPX-1, CPX1EC 3.4.17.-, CPXM, CPXM1, EC 3.4.17, EC 3.4.17.10, EC 3.4.17.3, Metallocarboxypeptidase CPX-1, probable carboxypeptidase X1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Carboxypeptidase X1/CPXM1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse CPXM is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CPXM1
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase X1/CPXM1 Ala21-Asp734 Accession # Q96SM3
Antigen Affinity-purified
RUO
Primary
56265
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only