missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase B1/CPB1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2897

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457854

  • £362.60 / 100µg

Estimated Shipment
10-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Carboxypeptidase B1/CPB1 Polyclonal antibody AF2897 Specifically detects Carboxypeptidase B1/CPB1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Carboxypeptidase B1/CPB1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P15086
carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, CPB1, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Carboxypeptidase B1/CPB1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) CPB1 is observed and less than 5% cross-reactivity with recombinant human (rh) CPA1, rhCPA2, and rmCPA4 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CPB1
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase B1/CPB1 His16-Tyr417 Accession # P15086
Immunogen affinity purified
RUO
Primary
1360
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only