missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase A2/CPA2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2896

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447854

  • £362.60 / 100µg

Estimated Shipment date
from Supplier 06-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Carboxypeptidase A2/CPA2 Polyclonal antibody AF2896 Specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Carboxypeptidase A2/CPA2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Affinity Purified
CPA2
NS0-derived rhCPA2 (aa 17 - 417) aa 17-419
Antigen Affinity-purified
RUO
Primary
1358
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), CPA2, EC 3.4.17, EC 3.4.17.15
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CPA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) CPA1 is observed and less than 1% cross-reactivity with rhCPB1 and recombinant mouse CPA4 is observed.
Human
Videos
Documents

For Research Use Only