Learn More
Carbohydrate Sulfotransferase 15/CHST15 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3365
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488124
  • £377.53 / 100µg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Carbohydrate Sulfotransferase 15/CHST15 Polyclonal antibody AF3365 Specifically detects Carbohydrate Sulfotransferase 15/CHST15 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
Carbohydrate Sulfotransferase 15/CHST15
Unconjugated
Q7LFX5
Goat
IgG
RUO
Polyclonal
Human
CyTOF, Flow Cytometry, Western Blotting
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human Carbohydrate Sulfotransferase 15/CHST15, Ser99-Thr561, Accession No. Q7LFX5
100μg
Primary
51363