missing translation for 'onlineSavingsMsg'
Learn More

C1qTNF1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3145

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458004

  • £377.53 / 100µg


Estimated Shipment
03-12-2018
Add to basket

Description

Description

C1qTNF1 Polyclonal antibody AF3145 Specifically detects C1qTNF1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

C1qTNF1/CTRP1
LYOPH
Western Blot 0.1 ug/mL
Q9BXJ1
C1q and tumor necrosis factor related protein 1, C1qTNF1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human CTRP1/C1qTNF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CTRP4 and rhCTRP5 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
C1QTNF1
Mouse myeloma cell line NS0-derived recombinant human CTRP1/C1qTNF1, Arg26-Pro281, Accession # Q9BXJ1
Antigen Affinity-purified
RUO
Primary
114897
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only