Learn More
BMP-9 Propeptide Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3879
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448424
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 24-02-2020
Description
Description
BMP-9 Propeptide Polyclonal antibody AF3879 Specifically detects BMP-9 Propeptide in Human samples. It is validated for Western Blotting
Specifications
Specifications
BMP-9
LYOPH
Western Blot 0.1 ug/mL
Q9UK05
BMP9, BMP9BMP-9Bone morphogenetic protein 9, GDF2, GDF-2, growth differentiation factor 2, growth/differentiation factor 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human BMP-9 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with mature recombinant human BMP-9 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GDF2
Chinese hamster ovary cell line CHO-derived recombinant human BMP-9 Propeptide Lys23-Arg319 Accession # Q9UK05
Immunogen affinity purified
RUO
Primary
2658
Reconstitute at 0.2 mg/mL in sterile PBS.