missing translation for 'onlineSavingsMsg'
Learn More

beta IG-H3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2935

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467874

  • £362.60 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

beta IG-H3 Polyclonal antibody AF2935 Specifically detects beta IG-H3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Beta Ig-h3/TGFBI
LYOPH
Western Blot 0.1 ug/mL
Q15582
beta IGH3, Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, TGFBIP, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human beta IG-H3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse beta IG-H3 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TGFBI
Mouse myeloma cell line NS0-derived recombinant human beta IG-H3 Gly24-His683 Accession # Q15582
Immunogen affinity purified
RUO
Primary
7045
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only