missing translation for 'onlineSavingsMsg'
Learn More

beta-Defensin 3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4435

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488724

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

beta-Defensin 3 Polyclonal antibody AF4435 Specifically detects beta-Defensin 3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

beta-Defensin 3
LYOPH
Western Blot 0.1 ug/mL
P81534
BD14, BD3, BD-3, beta-defensin 103, betaDefensin 3, Beta-defensin 3, DEFB103, DEFB103A, DEFB103B, DEFB3, DEFB-3, DEFB3DEFB103A, Defensin, beta 103, defensin, beta 103B, Defensin-like protein, hBD-3, HBD3HBP3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human beta-Defensin 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human beta-Defensin 2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DEFB103B
E. coli-derived recombinant human beta-Defensin 3, Gly23-Lys67, Accession # P81534
Immunogen affinity purified
RUO
Primary
55894
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only