missing translation for 'onlineSavingsMsg'
Learn More

beta-Arrestin 1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4560

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468794

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

beta-Arrestin 1 Polyclonal antibody AF4560 Specifically detects beta-Arrestin 1 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

beta-Arrestin 1
LYOPH
Immunohistochemistry 5-15 ug/mL
P49407
ARB1, ARR1, ARRB1, arrestin 2, Arrestin beta-1, arrestin, beta 1, betaArrestin 1, beta-arrestin-1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human beta-Arrestin 1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human beta-Arrestin 2 is observed.
Human
Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ARRB1
E. coli-derived recombinant human beta-Arrestin 1 Met1-Arg418 Accession # P49407
Immunogen affinity purified
RUO
Primary
408
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only