missing translation for 'onlineSavingsMsg'
Learn More

BACE-2 Ectodomain Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4097

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448554

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BACE-2 Ectodomain Polyclonal antibody AF4097 Specifically detects BACE-2 Ectodomain in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

BACE-2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q9Y5Z0
AEPLC, Asp 1, ASP1, ASP21, Aspartic-like protease 56 kDa, Aspartyl protease 1, BACE2, beta secretase 2, beta-secretase 2, beta-site amyloid beta A4 precursor protein-cleaving enzyme 2, Beta-site amyloid precursor protein cleaving enzyme 2, Beta-site APP cleaving enzyme 2, beta-site APP-cleaving enzyme 2, beta-site APP-cleaving enzyme 2, EC 3.4.2310ALP5656 kDa aspartic-like protease, CEAP1, Down region aspartic protease, Down syndrome region aspartic protease, DRAPBAE2, EC 3.4.23, EC 3.4.23.45, Memapsin-1, Membrane-associated aspartic protease 1, Theta-secretase, transmembrane aspartic proteinase Asp1
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse BACE-2 Ectodomain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) BACE-1, rhCathepsin D, and rhCathepsin E is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
BACE2
Mouse myeloma cell line NS0-derived recombinant human BACE-2 Ectodomain Gly63-Pro466 Accession # Q9Y5Z0
Immunogen affinity purified
RUO
Primary
25825
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only