missing translation for 'onlineSavingsMsg'
Learn More

APRIL/TNFSF13 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF884

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460295

  • £322.09 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

APRIL/TNFSF13 Polyclonal antibody AF884 Specifically detects APRIL/TNFSF13 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

APRIL/TNFSF13
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Affinity Purified
TNFSF13
E. coli-derived recombinant human APRIL/TNFSF13
Antigen Affinity-purified
RUO
Primary
8741
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TNFSF13, TRDL1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human APRIL/TNFSF13 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) Fas Ligand, rhTNF-alpha , rhTRAIL, and rhTRANCE is observed.
Human
Videos

For Research Use Only