missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-like Protein 4/ANGPTL4 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3485

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448194

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Angiopoietin-like Protein 4/ANGPTL4 Polyclonal antibody AF3485 Specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

Angiopoietin-like Protein 4/ANGPTL4
LYOPH
Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Q9BY76
Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ANGPTL4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) ANGPTL3, rhAngiopoietin-1, -2, -3, and -4 is observed. In Western blots, approximately 1% cross-reactivity with rhANGPLT4 N-terminal Fragment is observed.
Human, Primate
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ANGPTL4
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 4/ANGPLT4, Leu165-Ser406, Accession # Q9BY76
Antigen Affinity-purified
RUO
Primary
51129
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only