missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-like Protein 3/ANGPTL3 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3829

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438394

  • £314.17 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Angiopoietin-like Protein 3/ANGPTL3 Polyclonal antibody AF3829 Specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Angiopoietin-like Protein 3/ANGPTL3
LYOPH
Western Blot 0.1 ug/mL
Q9Y5C1
AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Angiopoietin-like 3 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Angiopoietin-like 3 is observed and less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-like 1 and rhAngiopoietin-like 4 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ANGPTL3
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Angiopoietin-like 3 (R&D Systems, Catalog # 3829-AN) Ser17-Glu460 Accession # Q9Y5C1
Antigen Affinity-purified
RUO
Primary
27329
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only