missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-like Protein 1/ANGPTL1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2204

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467404

  • £362.60 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Angiopoietin-like Protein 1/ANGPTL1 Polyclonal antibody AF2204 Specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Angiopoietin-like Protein 1/ANGPTL1
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
O95841
ANG-3, ANG3angioarrestin, Angiopoietin like Protein 1, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, ANGPTL1, AngY, AngY1, ARP1, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Angiopoietin-like 1/ANGPTL1 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) Angiopoietin-4 is observed and less than 5% cross-reactivity with rhAngiopietin-1 and rhAngiopoietin-2 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ANGPTL1
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 1/ANGPTL1 Ala39-Asp491 Accession # O95841
Immunogen affinity purified
RUO
Primary
9068
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only