Learn More
Amnionless Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1860
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13467194
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Amnionless Polyclonal antibody AF1860 Specifically detects Amnionless in Human samples. It is validated for Western Blotting
Specifications
Specifications
Amnionless/AMN
LYOPH
Western Blot 0.1 ug/mL
Q9BXJ7
AMN, amnionless homolog (mouse), PRO1028, protein amnionless, UNQ513/PRO1028, visceral endoderm-specific type 1 transmembrane protein
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Amnionless in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AMN
Mouse myeloma cell line NS0-derived recombinant human Amnionless Val20-Ala358 Accession # Q9BXJ7
Antigen Affinity-purified
RUO
Primary
81693
Reconstitute at 0.2 mg/mL in sterile PBS.