missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase P2/XPNPEP2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2490

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457594

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Aminopeptidase P2/XPNPEP2 Polyclonal antibody AF2490 Specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Aminopeptidase P2/XPNPEP2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
AAB96394
Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, XPNPEP2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Aminopeptidase P2/XPNPEP2 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Aminopeptidase P2 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
XPNPEP2
Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase P2/XPNPEP2 His22-Ala650 Accession # AAB96394
Immunogen affinity purified
RUO
Primary
7512
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only